Phenol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene
M-Clarity™ quality level = MQ200
33517-100G 33517-1KG 33517-500G 33517-6X1KG
SIAL33517-100GEA 75.1 EUR
SIAL33517-100G SIAL33517-1KG SIAL33517-500G SIAL33517-6X1KG
Phenol, Sigma-Aldrich®
Phenol
Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.7
Melting Pt: 40.8
Density: 1.0722 g/cm³ (20 °C)
Flash Pt: 81
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
REACH: 01-2119471329-32
Merck Index: 12,07390