2,2'-Dipyridylamine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,2'-Iminodipyridine, Di(pyridin-2-yl)amine, Chlorpheniramine Related Compound B

M-Clarity™ quality level = MQ200

D216402-5G D216402-25G
SIALD216402-5GEA 126 EUR
SIALD216402-5G SIALD216402-25G
2,2'-Dipyridylamine, Sigma-Aldrich®
2,2'-Dipyridylamine
Formula: C₁₀H₉N₃
MW: 171.2 g/mol
MDL Number: MFCD00006247
CAS Number: 1202-34-2
EINECS: 214-864-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR