1,3-Diphenylacetone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone

W239704-5KG-K
SIALW239704-5KG-KEA 3040 EUR
SIALW239704-5KG-K
1,3-Diphenylacetone, Sigma-Aldrich®
1,3-Diphenylacetone
Formula: (C₆H₅)CH₂C(O)CH₂(C₆H₅)
MW: 210.28 g/mol
Boiling Pt: 330
Melting Pt: 30…34 °C
Density: 1.195 g/cm³ (0 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004795
CAS Number: 102-04-5
EINECS: 203-000-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR