α,α,α,α',α',α'-Hexafluoro-m-xylene, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
251186-100G 251186-25G
SIAL251186-100GEA 70.8 EUR
SIAL251186-100G SIAL251186-25G
α,α,α,α',α',α'-Hexafluoro-m-xylene, Sigma-Aldrich®
α,α,α,α',α',α'-Hexafluoro-m-xylene
Formula: C₈H₄F₆
Boiling Pt: 116.3 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.383 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000392
CAS Number: 402-31-3
EINECS: 206-939-4
UN: 1993
ADR: 3,III