2,4-Lutidine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,4-Dimethylpyridine
M-Clarity™ quality level = MQ100
L3609-100ML L3609-500ML L3609-5ML
SIALL3609-100MLEA 110 EUR
SIALL3609-100ML SIALL3609-500ML SIALL3609-5ML
2,4-Lutidine, Sigma-Aldrich®
2,4-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.93 g/cm³ (20 °C)
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00006337
CAS Number: 108-47-4
EINECS: 203-586-8
UN: 1992
ADR: 3,III