Burners, Teclu
Supplier: BOCHEM
7100 7110
BOCH7100EA 145 EUR
BOCH7100 BOCH7110
Burners, Teclu
Heaters Gas Burners
These burners have air regulation and reach a maximum temperature of 1300 °C.
Certifikace: DIN 30665