1-Chloro-5-(trimethylsilyl)-4-pentyne ≥97%
Supplier: Thermo Fisher Scientific
Warning
|
H53393.06
H53393.14
H53393.06EA
56.5
EUR
H53393.06
H53393.14
1-Chloro-5-(trimethylsilyl)-4-pentyne ≥97%
1-Chloro-5-(trimethylsilyl)-4-pentyne
Purity: 97%
CAS No.: 77113-48-5
Molecular Formula: C8H15ClSi
Formula Weight: 174.75
Flash Point: 65°(149°F)
Refractive Index: 1.4560
Boiling Point: 75-77°/21mm
MDL No.: MFCD00671353
Formula:
C₈H₁₅ClSi Boiling Pt: 75…77 °C (21 mmHg) Density: 0.977 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00671353 CAS Number: 77113-48-5 |