(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: (S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid

M-Clarity™ quality level = MQ200

318035-1G 318035-250MG
SIAL318035-1GEA 160 EUR
SIAL318035-1G SIAL318035-250MG
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, Sigma-Aldrich®
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
MDL Number: MFCD00064200
CAS Number: 17257-71-5
EINECS: 241-292-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR