4-Hydroxyphenylboronic acid pinacol ester, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 4-Hydroxybenzeneboronic acid pinacol ester
M-Clarity™ quality level = MQ100
522570-1G 522570-5G
SIAL522570-1GEA 83.7 EUR
SIAL522570-1G SIAL522570-5G
4-Hydroxyphenylboronic acid pinacol ester, Sigma-Aldrich®
4-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
MW: 220.08 g/mol
MDL Number: MFCD02093756
CAS Number: 269409-70-3