4-Iodophenylboronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Iodobenzeneboronic acid pinacol ester, 2-(4-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

M-Clarity™ quality level = MQ100

699470-1G 699470-5G
SIAL699470-1GEA 106 EUR
SIAL699470-1G SIAL699470-5G
4-Iodophenylboronic acid pinacol ester, Sigma-Aldrich®
4-Iodophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BIO₂
MW: 329.97 g/mol
MDL Number: MFCD09953499
CAS Number: 73852-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR