Dihydromyrcenol, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W516406-1KG W516406-SAMPLE
SIALW516406-1KGEA 110 EUR
SIALW516406-1KG SIALW516406-SAMPLE
Dihydromyrcenol, Sigma-Aldrich®
Dihydromyrcenol
Formula: C₁₀H₂₀O
CAS Number: 18479-58-8
EINECS: 242-362-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR