Storage canes for Cryovial® tubes
Supplier: Simport Scientific
SIMPT313EA 35.8 EUR
SIMPT313
Storage canes for Cryovial® tubes
Cryogenic Straws
These aluminium storage canes can store upto five 1,2 ml or 2 ml tubes in liquid nitrogen containers such as Dewar flasks.