Filter tips
Supplier: KARTELL
9629 9630 9631 9632 9633 9634 9637 9638 9639
KART9629EA 506 EUR
KART9629 KART9630 KART9631 KART9632 KART9633 KART9634 KART9637 KART9638 KART9639
Filter tips
Pipette Tips
These filter tips made from PP are designed for use in molecular biology, cell cultures, bacteriology, virology, immunology and radioimmunology. Hydrophobic PE filters act as a barrier against aerosol contamination, preventing cross-contamination of samples and micropipettes.
  • DNA-free, RNA-free, cadmium-free, non pyrogenic
  • Sterilised by gamma radiation, autoclavable at +121 °C for 20 minutes
  • Recommended for use in automatic Kartell micropipettes
  • Suitable for use with most micropipette brands available on the market