α-Bromo-4-fluorotoulene

Supplier: Thermo Scientific
Danger

Synonyms: α-Bromo-p-fluorotoulene, 4-Fluorobenzyl bromide, p-Fluorobenzyl bromide

SB01932.1 SB01932.10 SB01932.50
MAYBSB01932.1EA 0 EUR
MAYBSB01932.1 MAYBSB01932.10 MAYBSB01932.50
α-Bromo-4-fluorotoulene
α-Bromo-4-fluorotoulene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 85 °C (1013 hPa)
Density: 1.517 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000359
CAS Number: 459-46-1
EINECS: 207-291-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR