3-Bromo-α,α,α-trifluorotoluene ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-Bromo-3-(trifluoromethyl)benzene, 3-Bromobenzotrifluoride
A14724.14 A14724.22 A14724.36
A14724.14EA 33.6 EUR
A14724.14 A14724.22 A14724.36
3-Bromo-α,α,α-trifluorotoluene ≥98%
3-Bromo-α,α,α-trifluorotoluene
Purity: 98+% CAS No.: 401-78-5 Molecular Formula: C7H4BrF3 Formula Weight: 225.01 Flash Point: 59°(138°F) Refractive Index: 1.4730 Odour: Aromatic Boiling Point: 151-152° Beilstein Reference: 1449557 UN No.: 1993 MDL No.: MFCD00000380
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 151…152 °C
Melting Pt: 37.5…39 °C
Density: 1.613
Flash Pt: 59 °C (138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000380
CAS Number: 401-78-5
EINECS: 206-932-6
UN: 1993
ADR: 3 III