α,4-Dibromo-2-fluorotoluene ≥98%
Supplier: Thermo Scientific
Danger
Synonyms: 4-Bromo-1-(bromomethyl)-2-fluorobenzene, 4-Bromo-2-fluorobenzyl bromide
B21331.06 B21331.14 B21331.22
B21331.06EA 47.9 EUR
B21331.06 B21331.14 B21331.22
α,4-Dibromo-2-fluorotoluene ≥98%
α,4-Dibromo-2-fluorotoluene
Purity: 98% CAS No.: 76283-09-5 Molecular Formula: C7H5Br2F Formula Weight: 267.92 Melting Point: 33-37° Boiling Point: 126°/9mm UN No.: 3261 MDL No.: MFCD00055467
Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Boiling Pt: 126 °C (9 mmHg)
Melting Pt: 33…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00055467
CAS Number: 76283-09-5
EINECS: 278-412-7
UN: 3261
ADR: 8,II