4-Bromo-α,α,α-trifluorotoluene

Supplier: Thermo Scientific
Danger

Synonyms: 4-Bromobenzotrifluoride, 1-Bromo-4-(trifluoromethyl)benzene

SB01902.1 SB01902.10 SB01902.50
MAYBSB01902.1EA 0 EUR
MAYBSB01902.1 MAYBSB01902.10 MAYBSB01902.50
4-Bromo-α,α,α-trifluorotoluene
4-Bromo-α,α,α-trifluorotoluene
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 154 °C (1013 hPa)
Density: 1.6347 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000398
CAS Number: 402-43-7
EINECS: 206-943-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR