4-Tolualdehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: p-Tolualdehyde, p-Tolylaldehyde, p-Methylbenzaldehyd, 4-Methylbenzaldehyd, 4-Methylbenzaldehyde, 4-Tolylaldehyde
M-Clarity™ quality level = MQ100
T35602-500G T35602-100G T35602-5G
SIALT35602-500GEA 98.9 EUR
SIALT35602-500G SIALT35602-100G SIALT35602-5G
4-Tolualdehyde, Sigma-Aldrich®
4-Tolualdehyde
Formula: C₈H₈O
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –6 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006954
CAS Number: 104-87-0
EINECS: 203-246-9