Benzyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (Chloromethyl)benzene, Chlorophenylmethane, α-Chlorotoluene
M-Clarity™ quality level = MQ200
185558-50G 185558-1KG 185558-250G 185558-2KG
SIAL185558-50GEA 32.9 EUR
SIAL185558-50G SIAL185558-1KG SIAL185558-250G SIAL185558-2KG
Benzyl chloride, Sigma-Aldrich®
Benzyl chloride
Formula: (C₆H₅)CH₂Cl
MW: 126.59 g/mol
Boiling Pt: 179.3 °C (1013 hPa)
Melting Pt: –41.2 °C
Density: 1.103 g/cm³ (20 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00000889
CAS Number: 100-44-7
EINECS: 202-853-6
UN: 1738
ADR: 6.1,II
Merck Index: 13,01131