2,4-Dichlorotoluene 99%
Supplier: Thermo Scientific
Warning
Synonyms: 1,3-Dichloro-4-methylbenzene
163280010 163282500
ACRO163280010EA 134 EUR
ACRO163280010 ACRO163282500
2,4-Dichlorotoluene 99%
2,4-Dichlorotoluene
Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: –14 °C
Density: 1.24 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000583
CAS Number: 95-73-8
EINECS: 202-445-8
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
GC ≥98.5 %