α,3-Dichlorotoluene
Supplier: Thermo Scientific
Synonyms:
1-Chloro-3-(chloromethyl)benzene, 3-Chlorobenzyl chloride, 3-Chlorobenzylchloride
SB00676.1
SB00676.10
SB00676.50
MAYBSB00676.1EA
0
EUR
MAYBSB00676.1
MAYBSB00676.10
MAYBSB00676.50
α,3-Dichlorotoluene
α,3-Dichlorotoluene
Formula:
C₇H₆Cl₂ Boiling Pt: 216 °C (1013 hPa) Melting Pt: 69…71 °C Density: 1.27 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000905 CAS Number: 620-20-2 EINECS: 210-629-4 UN: 2235 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...