3,4-Dichloro-α,α,α-trifluorotoluene
Supplier: Thermo Scientific
Warning
Synonyms: 1,2-Dichloro-4-(trifluoromethyl)benzene, 3,4-Dichlorobenzotrifluoride
SB01092.1 SB01092.10 SB01092.50
MAYBSB01092.1EA 0 EUR
MAYBSB01092.1 MAYBSB01092.10 MAYBSB01092.50
3,4-Dichloro-α,α,α-trifluorotoluene
3,4-Dichloro-α,α,α-trifluorotoluene
Formula: C₇H₃Cl₂F₃
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –0.2 °C
Density: 1.486 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000555
CAS Number: 328-84-7
EINECS: 206-337-1
UN: 2810
ADR: 6.1,III