α-Chloro-3-fluorotoulene
Supplier: Thermo Scientific
Danger
Synonyms: 1-(Chloromethyl)-3-fluorobenzene, α-Chloro-m-fluorotoulene, 3-Fluorobenzyl chloride
SB01021.1 SB01021.10 SB01021.50
MAYBSB01021.1EA 0 EUR
MAYBSB01021.1 MAYBSB01021.10 MAYBSB01021.50
α-Chloro-3-fluorotoulene
α-Chloro-3-fluorotoulene
Formula: C₇H₆ClF
Boiling Pt: 67 °C (1013 hPa)
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000904
CAS Number: 456-42-8
EINECS: 207-264-8
UN: 2920
ADR: 8,II