α,2-Dichloro-6-fluorotoluene
Supplier: Thermo Scientific
Danger
Synonyms: 2-Chloro-6-fluorobenzyl chloride, 1-Chloro-2-(chloromethyl)-3-fluorobenzene
SB00677.1 SB00677.10 SB00677.50
MAYBSB00677.1EA 0 EUR
MAYBSB00677.1 MAYBSB00677.10 MAYBSB00677.50
α,2-Dichloro-6-fluorotoluene
α,2-Dichloro-6-fluorotoluene
Formula: C₇H₅Cl₂F
MW: 179.02 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 7 °C
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00000896
CAS Number: 55117-15-2
EINECS: 259-487-5
UN: 3265
ADR: 8,II