2-Chloro-6-fluorotoluene

Supplier: Thermo Scientific

SB00544.1 SB00544.10 SB00544.50
MAYBSB00544.1EA 0 EUR
MAYBSB00544.1 MAYBSB00544.10 MAYBSB00544.50
2-Chloro-6-fluorotoluene
2-Chloro-6-fluorotoluene
Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 154 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000570
CAS Number: 443-83-4
EINECS: 207-141-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR