α,α,α,4-Tetrafluorotoluene ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Fluorobenzotrifluoride, 1-Fluoro-4-(trifluoromethyl)benzene
A13377.06
A13377.14
A13377.06EA
55.1
EUR
A13377.06
A13377.14
α,α,α,4-Tetrafluorotoluene ≥98%
α,α,α,4-Tetrafluorotoluene
Purity: 98+%
CAS No.: 402-44-8
Molecular Formula: C7H4F4
Formula Weight: 164.10
Flash Point: 10°(50°F)
Refractive Index: 1.4010
Melting Point: -42°
Boiling Point: 103-105°
Beilstein Reference: 1909390
UN No.: 1993
MDL No.: MFCD00000399
Formula:
C₇H₄F₄ MW: 164.1 g/mol Boiling Pt: 103…105 °C Melting Pt: –42 °C Density: 1.295 Flash Pt: 10 °C (50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000399 CAS Number: 402-44-8 EINECS: 206-944-1 UN: 1993 ADR: 3,II |