2-Fluoro-5-nitrobenzotrifluoride

Supplier: Thermo Scientific
Warning

Synonyms: 2-Fluoro-5-nitrobenzotrifluoride, 1-Fluoro-4-nitro-2-(trifluoromethyl)benzene

SB01728.1 SB01728.10 SB01728.50
MAYBSB01728.1EA 0 EUR
MAYBSB01728.1 MAYBSB01728.10 MAYBSB01728.50
2-Fluoro-5-nitrobenzotrifluoride
2-Fluoro-5-nitrobenzotrifluoride
Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Boiling Pt: 99…110 °C (35 mmHg)
Melting Pt: 22…24 °C
Density: 1.52 g/cm³ (20 °C)
MDL Number: MFCD00024576
CAS Number: 400-74-8
EINECS: 206-925-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR