Clips assortment for conical joints, metal, KECK™, DURAN®
Supplier: DWK Life Sciences
KECK™
SCOT290330008EA 79.81 EUR
SCOT290330008
Clips assortment for conical joints, metal, KECK™, DURAN®
Clamps NS Joint Clamps
Metal.
Delivery information: The set includes the following clips: 2 × KCM 14, 2 × KCM 19, 1 × KCM 29, 1 × KCM 24.