3-Ethylaniline 98%
Supplier: Thermo Fisher Scientific
Danger
174550100 174550500 174552500
ACRO174550100EA 49.7 EUR
ACRO174550100 ACRO174550500 ACRO174552500
3-Ethylaniline 98%
3-Ethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 212
Melting Pt: –8 °C
Density: 0.97 g/cm³
Flash Pt: 85
Storage Temperature: Ambient
MDL Number: MFCD00007818
CAS Number: 587-02-0
EINECS: 209-594-8
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5540 to 1.5580 (20°C, 589 nm)
GC ≥97.5 %