4-Nitrotoluene 99%
Supplier: Thermo Fisher Scientific
Danger
129050010 129050050 129051000
ACRO129050010EA 80.2 EUR
ACRO129050010 ACRO129050050 ACRO129051000
4-Nitrotoluene 99%
4-Nitrotoluene
Formula: C₇H₇NO₂
Boiling Pt: 238.3 °C (1013 hPa)
Melting Pt: 51…54 °C
Density: 1,39 g/cm³ (20 °C)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00007366
CAS Number: 99-99-0
EINECS: 202-808-0
UN: 3446
ADR: 6.1,II
Merck Index: 12,06748
Order Now
Specification Test Results
Appearance Yellow Crystalline solid
Infrared spectrum Conforms
Melting point 51 to 54 °C
GC ≥98.5 %