4-Nitrotoluene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
N27322-100G
N27322-1KG
SIALN27322-100GEA
33.5
EUR
SIALN27322-100G
SIALN27322-1KG
4-Nitrotoluene, Sigma-Aldrich®
4-Nitrotoluene
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 238.3 Melting Pt: 51 Density: 1.39 g/cm³ (20 °C) Flash Pt: 106 Storage Temperature: Ambient |
MDL Number:
MFCD00007366 CAS Number: 99-99-0 EINECS: 202-808-0 UN: 3446 ADR: 6.1 II Merck Index: 12,06748 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...