4-tert-Butylphenol 97%
Supplier: Thermo Scientific
Danger
108000010 108000025 108000050
ACRO108000010EA 45.6 EUR
ACRO108000010 ACRO108000025 ACRO108000050
4-tert-Butylphenol 97%
4-tert-Butylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 96…100 °C
Density: 0.9 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002367
CAS Number: 98-54-4
EINECS: 202-679-0
UN: 3077
ADR: 9,III
Merck Index: 13,01584
Order Now
Specification Test Results
Appearance White to light beige Flakes or pastilles
Infrared spectrum Conforms
Melting point 96 to 100 °C
Water ≤0.5 % (K.F.)
GC ≥96.0 %