4-tert-Butylphenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
B99901-100G B99901-1KG B99901-5G
SIALB99901-100GEA 49.5 EUR
SIALB99901-100G SIALB99901-1KG SIALB99901-5G
4-tert-Butylphenol, Sigma-Aldrich®
4-tert-Butylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 95…98 °C
Density: 1.017 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002367
CAS Number: 98-54-4
EINECS: 202-679-0
UN: 2430
ADR: 8,III
Merck Index: 13,01584