2-Ethylphenol

Supplier: EHRENSTORFER GMBH

EHERC13350000EA 35 EUR
EHERC13350000
2-Ethylphenol
2-Ethylphenol
Formula: C₈H₁₀O
MW: 122,17 g/mol
Boiling Pt: 204,5 °C (1013 hPa)
Melting Pt: –18 °C
Density: 1,0371 g/cm³ (0 °C)
MDL Number: MFCD00002249
UN: 3145
ADR: 8,III
Merck Index: 13,03871

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR