Ethylbenzene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Phenylethane
M-Clarity™ quality level = MQ100
296848-100ML 296848-1L
SIAL296848-100MLEA 112 EUR
SIAL296848-100ML SIAL296848-1L
Ethylbenzene, Sigma-Aldrich®
Ethylbenzene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 136.2 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS Number: 100-41-4
EINECS: 202-849-4
UN: 1175
ADR: 3,II
Merck Index: 13,03801
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67d12bb66494ff09657a029fa1e68b09af34358&vid=3e5678dd6a0342c5ec35b93e48587e8125977cf7&prid=web&clid=SAPDAM