4-Chloro-7-nitrobenzofurazan 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: NBD chloride, 4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole, NBD-Cl
172390010 172390050 172390250
ACRO172390010EA 29.9 EUR
ACRO172390010 ACRO172390050 ACRO172390250
4-Chloro-7-nitrobenzofurazan 98%
4-Chloro-7-nitrobenzofurazan
Formula: C₆H₂ClN₃O₃
MW: 199.55 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00005808
CAS Number: 10199-89-0
EINECS: 233-496-4
Order Now
Specification Test Results
Appearance Yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 99 °C
Sulfated ash ≤0.1 %
GC ≥97.5 %