1,3-Dicyclohexylurea ≥98%

Supplier: Thermo Scientific

Synonyms: N,N′-Dicyclohexylurea, N,N'-Dicyclohexylurea

B20805.14 B20805.22 B20805.36
B20805.14EA 55 EUR
B20805.14 B20805.22 B20805.36
1,3-Dicyclohexylurea ≥98%
1,3-Dicyclohexylurea

Purity: 98% CAS No.: 2387-23-7 Molecular Formula: C13H24N2O Formula Weight: 224.35 Melting Point: 230-233° MDL No.: MFCD00003829

Formula: C₁₃H₂₄N₂O
MW: 224.35 g/mol
Melting Pt: 230…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00003829
CAS Number: 2387-23-7
EINECS: 219-213-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR