Zinc chloride 0.7 M in THF
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
H32962.AE H32962.AP
H32962.AEEA 136 EUR
H32962.AE H32962.AP
Zinc chloride 0.7 M in THF
Zinc chloride
CAS No.: 7646-85-7 Molecular Formula: Cl2Zn Formula Weight: 136.29 UN No.: 1993 MDL No.: MFCD00011295
Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732
Melting Pt: 283
Density: 0.948
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 1993
ADR: 3 II
Merck Index: 13,10185