Methyl-3-chlorothiophene-2-carboxylate

Supplier: Thermo Scientific

GK01295DA GK01295EA
MAYBGK01295DAEA 0 EUR
MAYBGK01295DA MAYBGK01295EA
Methyl-3-chlorothiophene-2-carboxylate
Methyl 3-chlorothiophene-2-carboxylate
Formula: C₆H₅ClO₂S
MDL Number: MFCD00068135
CAS Number: 88105-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR