4'-Aminoacetophenone oxime

Supplier: Thermo Scientific

S11428.1 S11428.10 S11428.50
MAYBS11428.1EA 0 EUR
MAYBS11428.1 MAYBS11428.10 MAYBS11428.50
4'-Aminoacetophenone oxime
4'-Aminoacetophenone oxime
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
CAS Number: 38063-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR