tert-Butylchlorodimethylsilane
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride
SB01935.1
SB01935.10
SB01935.50
MAYBSB01935.1EA
0
EUR
MAYBSB01935.1
MAYBSB01935.10
MAYBSB01935.50
tert-Butylchlorodimethylsilane
tert-Butylchlorodimethylsilane
Formula:
C₆H₁₅ClSi MW: 150.72 g/mol Boiling Pt: 125 °C (1013 hPa) Melting Pt: 87…90 °C Density: 0,81 g/cm³ (20 °C) Flash Pt: 22 °C |
MDL Number:
MFCD00000501 CAS Number: 18162-48-6 EINECS: 242-042-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...