2'-Aminoacetophenone oxime

Supplier: Thermo Scientific

SEW04822.250 SEW04822.1
MAYBSEW04822.250EA 0 EUR
MAYBSEW04822.250 MAYBSEW04822.1
2'-Aminoacetophenone oxime
2'-Aminoacetophenone oxime
Formula: C₈H₁₀N₂O
CAS Number: 4964-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR