(S,S)-(-)-Hydrobenzoin, Sigma-Aldrich®
Supplier: Merck
Synonyms: (1S,2S)-1,2-Diphenylethane-1,2-diol
256269-25G 256269-5G
SIAL256269-25GEA 273 EUR
SIAL256269-25G SIAL256269-5G
(S,S)-(-)-Hydrobenzoin, Sigma-Aldrich®
(S,S)-(-)-Hydrobenzoin
Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
MDL Number: MFCD00064255
CAS Number: 2325-10-2