N-Methylmorpholine-N-oxide 50% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Methylmorpholine N-Oxide, 4-Methylmorpholine 4-oxide
M-Clarity™ quality level = MQ200
258822-100G 258822-1KG
SIAL258822-100GEA 86.6 EUR
SIAL258822-100G SIAL258822-1KG
N-Methylmorpholine-N-oxide 50% in aqueous solution, Sigma-Aldrich®
N-Methylmorpholine N-oxide
Formula: C₅H₁₁NO₂
MDL Number: MFCD00005947
CAS Number: 7529-22-8
EINECS: 231-391-8