2-Butanone oxime, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
332828-1L 332828-25ML
SIAL332828-1LEA 82.2 EUR
SIAL332828-1L SIAL332828-25ML
2-Butanone oxime, Sigma-Aldrich®
2-Butanone oxime
Formula: C₄H₉NO
MDL Number: MFCD00013935
CAS Number: 96-29-7
EINECS: 202-496-6