4-Aminobenzylamine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-(Aminomethyl)aniline, alpha-Amino-p-toluidine

M-Clarity™ quality level = MQ100

368466-10G 368466-50G
SIAL368466-10GEA 88.3 EUR
SIAL368466-10G SIAL368466-50G
4-Aminobenzylamine, Sigma-Aldrich®
4-Aminobenzylamine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00075513
CAS Number: 4403-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR