(±)-α-Ethylbenzylamine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

448680-10G 448680-1G
SIAL448680-10GEA 205 EUR
SIAL448680-10G SIAL448680-1G
(±)-α-Ethylbenzylamine, Sigma-Aldrich®
(±)-α-Ethylbenzylamine
Formula: C₉H₁₃N
MW: 135.21 g/mol
CAS Number: 2941-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR