1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers), Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,4-Cyclohexanedicarboxylic acid
M-Clarity™ quality level = MQ200
538035-100G 538035-1KG
SIAL538035-100GEA 55.4 EUR
SIAL538035-100G SIAL538035-1KG
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers), Sigma-Aldrich®
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)
Formula: C₈H₁₂O₄
MW: 172.18 g/mol
MDL Number: MFCD00001465
CAS Number: 1076-97-7
EINECS: 214-068-6