Acetophenone oxime, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methyl phenyl ketoxime, 1-Phenyl-1-ethanone oxime, Acetophenone Oxime
M-Clarity™ quality level = MQ100
647659-25G 647659-5G
SIAL647659-25GEA 245 EUR
SIAL647659-25G SIAL647659-5G
Acetophenone oxime, Sigma-Aldrich®
Acetophenone oxime
Formula: C₈H₉NO
MW: 135.17 g/mol
MDL Number: MFCD00013931
CAS Number: 613-91-2
EINECS: 210-360-2