3,4-Dimethoxyphenethyl bromide, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-(2-Bromoethyl)-1,2-dimethoxybenzene
653675-1G
653675-5G
SIAL653675-1GEA
88.1
EUR
SIAL653675-1G
SIAL653675-5G
3,4-Dimethoxyphenethyl bromide, Sigma-Aldrich®
3,4-Dimethoxyphenethyl bromide
Formula:
C₁₀H₁₃BrO₂ MW: 245.12 g/mol |
CAS Number:
40173-90-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...